Skip to Content
MilliporeSigma

R4530

Anti-Rab21 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RAB21, member RAS oncogene family

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~25 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using whole extract of human HeLa cells, indirect immunofluorescence: 5-10 μg/mL using mouse 3T3 and rat NRK cells fixed and permeabilized with 4% paraformaldehyde followed by 0.4% saponin, western blot: 5-10 μg/mL using whole extract of human A431 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB21(23011)
mouse ... Rab21(216344)
rat ... Rab21(299799)

General description

Ras-related protein Rab-21 (Rab21), a close homolog of Rab5, is ubiquitously expressed in human tissues. It localizes mainly to the early endocytic pathway. Rab21 is mapped to human chromosome 12q21.1 It belongs to the Rab family of small guanosine triphosphatases (GTPases). Rab proteins contain conserved regions, C-terminal hypervariable domains with a cysteine motif.

Immunogen

synthetic peptide corresponding to amino acid residues 203-216 of human RAB21, with an added cysteine, conjugated to KLH. The corresponding sequence differs by one amino acid in rat and mouse.

Application

Anti-Rab21 (C-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Anti-Rab21 (C-terminal) recognizes human, rat, and mouse Rab21.
Ras-related protein Rab-21 (Rab21) and Rab5 are integrin-associated proteins, acting as positive regulators of integrin traffic. The association of Rab21 with integrins modulates cell adhesion and migration. It is also involved in endosome morphology and function. Rab21 is associated with squamous carcinoma and its expression is elevated in glioma.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

For continuous use, store at 2-8 °C for up to one month. For extendedstorage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library