Skip to Content
MilliporeSigma

SML4136

BB2-50F

≥98% (HPLC)

Synonym(s):

3-Amino-N-(aminoiminomethyl)-6-(2-benzofuranyl)-5-(hexahydro-1H-azepin-1-yl)-2-pyrazinecarboxamide, 3-Amino-5-(azepan-1-yl)-6-(benzofuran-2-yl)-N-carbamimidoylpyrazine-2-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H23N7O2
CAS Number:
Molecular Weight:
393.44
NACRES:
NA.21
UNSPSC Code:
12352203
eCl@ss:
32150313
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(NC(C1=NC(C2=CC(C=CC=C3)=C3O2)=C(N4CCCCCC4)N=C1N)=O)=N

Biochem/physiol Actions

Cell penetrant, selective and potent multi-targeting bioenergetic inhibitor of succinate dehydrogenase and the F1Fo-ATP synthase in Mycobacterium tuberculosis; bactericidal antibiotic in MTB.
BB2-50F, an amiloride derivative, is a cell penetrant, selective and potent multi-targeting bioenergetic inhibitor of succinate dehydrogenase and the F1Fo-ATP synthase in Mycobacterium tuberculosis. BB2-50F act as fast acting bactericidal antibiotic in both replicating and nonreplicating cultures of M. tuberculosis (MIC = 2.6 µM in M. tuberculosis mc2 6230). BB2-50F is a potent inhibitor of bacterial motility.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library