Product Name
Pyridine analytical standard
Product Number
02486
Product Brand
sial
CAS Number
110-86-1
Molecular Weight
79.10
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| PURITY (GC AREA %) | ≥ 99.9 % |
| REFRACTIVE INDEX N20/D | 1.509 - 1.511 |
| WATER (COULOMETR.) | ≤ 0.05 % |
| EXPIRY PERIOD | 36 MONTHS |