Product Name
Triclopyr PESTANAL®, analytical standard
Product Number
32016
Product Brand
sial
CAS Number
55335-06-3
Molecular Weight
256.47
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT147 - 154 C
WATER≤ 3.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS