Product Name
Pirbuterol acetate VETRANAL®, analytical standard
Product Number
32142
Product Brand
sial
CAS Number
65652-44-0
Molecular Weight
300.35
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 99.0 %
MELTING POINTFOR INFORMATION ONLY
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RESIDUAL SOLVENTS≤ 1.0 %
EXPIRY PERIOD60 MONTHS