Product Name
Metolachlor, predominantly S-Metolachlor PESTANAL®, analytical standard
Product Number
33859
Product Brand
sial
CAS Number
87392-12-9
Molecular Weight
283.79
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
Enantiomer Content (HPLC)≥ 60 % S-Metolachlor
SPECIFIC ROTATION (20/D)-8.50 - -5.50 Degrees
CONCENTRATIONC=2 in HEXANE
REFRACTIVE INDEX N20/D1.5280 - 1.5320
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS