Product Name
Didecyl phthalate analytical standard
Product Number
36933
Product Brand
sial
CAS Number
84-77-5
Molecular Weight
446.66
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.4795 - 1.4835
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS