Product Name
Azamethiphos PESTANAL®, analytical standard
Product Number
45331
Product Brand
sial
CAS Number
35575-96-3
Molecular Weight
324.68
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT88 - 93 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS