Product Name
Tepraloxydim PESTANAL®, analytical standard
Product Number
46331
Product Brand
sial
CAS Number
149979-41-9
Molecular Weight
341.83
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Faint Beige to Beige and Light Brown
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0% (sum of isomers)
MELTING POINT71 - 77 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS