Product Name
8-Aminonaphthalene-1,3,6-trisulfonic acid disodium salt BioReagent, suitable for fluorescence, ≥90% (CE)
Product Number
08658
Product Brand
sigma
CAS Number
5398-34-5
Molecular Weight
427.34
Test
Specification
APPEARANCE (COLOR)White to Light Brown
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 90.0 %
WATER6 - 10 %
CARBON 13 NMR SPECTRUMCONFORMS TO STRUCTURE
LUMINESCENCECORRESPONDS TO REQUIREMENTS
EMISSION MAXIMUM512 ± 5 nm