
4-Propylcyclohexanone

Synonym(s):
4-Propylcyclohexanone
Empirical Formula (Hill Notation):
C9H16O
CAS Number:
Molecular Weight:
140.22
Products
Brand
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing