Skip to Content
MilliporeSigma
M16 Medium With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered

M16 Medium

M16 Medium With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered
Synonym(s):
Cell Culture Medium, Cell Culture Medium M16, M16 Medium Solution

Products

Product Category
Brand
Physical Form
Quality Segment
Technique