Skip to Content
MilliporeSigma
MultiScreen Column Loader 45µL, Black, Non-Sterile

MultiScreen Column Loader

MultiScreen Column Loader 45µL, Black, Non-Sterile
Synonym(s):
Column Loader

Products

Product Category
Brand
Product Line
Wells
Quality Segment
Application
Sterility
Technique