
N-Methyl Lenalidomide

Synonym(s):
N-Methyl Lenalidomide, (3-(4-(methylamino)-1-oxoisoindolin-2-yl)piperidine-2,6-dione)
Empirical Formula (Hill Notation):
C14H15N3O3
CAS Number:
Molecular Weight:
273.29
Products
Product Category
Manufacturer
Quality Segment
Application
Special Grade
Showing 1-2 of 2