Skip to Content
Merck

BDPH9C021CA2

Bide Pharmatech

(4R,4'R)-2,2'-(2,3-Dihydro-1H-indene-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Synonym(s):

(4R,4'R)-2,2'-(2,3-dihydro-1H-indene-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C29H28O2N2
CAS Number:
Molecular Weight:
436.55
MDL number:
Product BDPH9C021CA2 is not currently sold in your country.


Inchi Code

1S/C29H28N2O2/c1-3-9-21(10-4-1)15-25-19-32-27(30-25)29(17-23-13-7-8-14-24(23)18-29)28-31-26(20-33-28)16-22-11-5-2-6-12-22/h1-14,25-26H,15-20H2/t25-,26-/m1/s1

Inchi Key

UMTCVPCXPUJJIQ-CLJLJLNGSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid or lump



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library