Quality Level
usage
500-1428 purifications
sustainability
Greener Alternative Product
concentration
45-55 mg/mL (Magnetic particle)
test parameters
total test time: 5 min
suitability
suitable for nucleic acid purification
greener alternative category
storage temp.
2-30°C
General description
Ultra pure MagPrep® Magnetic Particles are designed for efficient RNA and DNA isolation in both manual and high-throughput automated platforms. The highly versatile silica magnetic particles are compatible with most standard buffer formulations for silica-based nucleic acid purification, and in particular can be paired with specially-formulated Ultra pure MagPrep® buffers for high, clean yield free of nuclease and DNA contaminants. The small particle size, less than 100 nm, allows for rapid magnetic clearance and easy handling. Ultra pure MagPrep® Magnetic Particles are quality tested to be free of contaminant nucleases and DNA, reducing the likelihood of false positives or negatives in downstream assays.For research or further manufacturing use only. Not intended for direct use in humans or animals. Non-TSCA use only in US.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.
Application
- Nucleic Acid Isolation
- Manual or high-throughput automated isolations
- RT-qPCR
- Digital PCR
- Biological samples can be used
- Room temperature stable reagents
- DNA free
- DNase free
- RNase free
- NICKase free
- simple and fast protocols
Other Notes
- UPMPPART-20ML - Usage sufficient for ~1000-1,428 purifications
- UPMPPART-10ML - Usage sufficient for ~500-715 purifications
Legal Information
MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany
Kit Components Only
Product No.
Description
- UPMPPART-2ML-KC Ultra pure MagPrep Magnetic Particles
- UPMPCFDBB-50ML-KC Ultra pure MagPrep cfDNA Binding Buffer
- UPMPWB1-100ML-KC Ultra pure MagPrep Wash Buffer 1
- UPMPWB2-100ML-KC Ultra pure MagPrep Wash Buffer 2
- UPMPEB-10ML-KC Ultra pure MagPrep Elution Buffer
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service