Skip to Content
Merck

V900116

Sucrose

99% (GC), Vetec, reagent grade

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
EC Number:
200-334-9
UNSPSC Code:
12161700
PubChem Substance ID:
Beilstein/REAXYS Number:
90825
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Sucrose, 99% (GC), Vetec, reagent grade

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

grade

reagent grade

product line

Vetec

assay

99% (GC)

form

crystals

technique(s)

gas chromatography (GC): suitable

mp

185-187 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Sucrose can be used:
  • As a component of culture media for microorganisms.
  • As an impermeant solute in the study of forward osmosis membrane parameters.

General description

Sucrose is a disaccharide (glucose and fructose), which is commonly known as table sugar. It is a non-reducing sugar abundantly found in nature. It is present as the main constituent of soft drinks, beverages, and many food products.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antifungal properties of novel N-and α,β-substituted succinimides against dermatophytes
Lopez, S N, et al.
Arzneimittelforschung, 53(04), 280-288 (2003)
Sucrose-A prospering and sustainable organic raw material
Carbohydrates in Sustainable Development (2010)
Understanding membrane parameters of a forward osmosis membrane based on nonequilibrium thermodynamics
Fang Y, et al.
Journal of Membrane Science , 437(04), 72-81 (2013)
The structure of sucrose in the crystal and in solution
Sucrose: Properties and Applications (1995)
Denise Harrison et al.
Pediatrics, 130(5), 918-925 (2012-10-10)
The use of oral sucrose has been the most extensively studied pain intervention in newborn care to date. More than 150 published studies relating to sweet-taste-induced calming and analgesia in human infants have been identified, of which 100 (65%) include

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service