Merck
Novel HDL-based therapeutic agents.
Pharmacology & therapeutics (2012-04-03)
Tisha R Joy
ABSTRACT
Reduction in low-density lipoprotein cholesterol (LDL-C) levels has been associated with a 25-30% reduction in cardiovascular disease risk. However, there still remains a significant and quantifiable risk. Since epidemiologic data have demonstrated that low levels of high-density lipoprotein cholesterol (HDL-C) are associated with an increased risk for cardiovascular disease, novel therapeutic agents are currently being developed to either raise HDL-C levels or enhance HDL functionality. This review will highlight some of these therapeutic agents including cholesteryl ester transfer protein inhibitors, apolipoprotein A-I mimetics, RVX-208, and apolipoprotein A-I based infusion therapies. For each therapeutic class, an overview of the mechanism of action, pharmacokinetic data, and efficacy/safety evidence will be provided.
Sigma-Aldrich
Lipoprotein, high density from human plasma, ≥95% (SDS-PAGE), solution
Pricing and availability is not currently available.