Merck
  • Inhibitory effects of novel synthetic methimazole derivatives on mushroom tyrosinase and melanogenesis.
Inhibitory effects of novel synthetic methimazole derivatives on mushroom tyrosinase and melanogenesis.
Bioorganic & medicinal chemistry (2014-04-08)
Chin-Feng Chan, Shih-Ting Lai, Yi-Cin Guo, Ming-Jen Chen
ABSTRACT
In this study, we synthesized 4 methimazole (2-mercapto-1-methylimidazole, MMI) derivatives. The kinetics of inhibition on mushroom tyrosinase by methimazole and its derivatives were investigated. The results indicated that tert-butyl 3-methyl-2-sulfanylidene-2,3-dihydro-1H-imidazole-1-carboxylate (compound 3; 3), 2-mercaptoimidazole (MI; compound 1; 1) and MMI (compound 2; 2) significantly inhibited tyrosinase activity in a dose-dependent manner, exhibiting an IC50 value of 1.50mM, 4.11 mM, and 1.43 mM. However, compound 4 (4), compound 5 (5), and compound 6 (6) exerted no inhibitory effect on mushroom tyrosinase activity. Kinetic analysis indicated that 3 was a noncompetitive tyrosinase inhibitor, whereas both 1 and 2 were exhibited as mixed-type tyrosinase inhibitors. Furthermore, 3 exerted a potent inhibitory effect on intracellular melanin formation in the B16/F10 murine melanoma cells and did not cause cytotoxicity, as 1 and 2 did.
Sigma-Aldrich
2-Mercaptoimidazole, 98%
Pricing and availability is not currently available.