Merck
  • Dissection of mitochondrial Ca2+ uptake and release fluxes in situ after depolarization-evoked [Ca2+](i) elevations in sympathetic neurons.
Dissection of mitochondrial Ca2+ uptake and release fluxes in situ after depolarization-evoked [Ca2+](i) elevations in sympathetic neurons.
The Journal of general physiology (2000-02-29)
S L Colegrove, M A Albrecht, D D Friel
ABSTRACT
We studied how mitochondrial Ca2+ transport influences [Ca2+](i) dynamics in sympathetic neurons. Cells were treated with thapsigargin to inhibit Ca2+ accumulation by SERCA pumps and depolarized to elevate [Ca2+(i); the recovery that followed repolarization was then examined. The total Ca2+ flux responsible for the [Ca2+](i) recovery was separated into mitochondrial and nonmitochondrial components based on sensitivity to the proton ionophore FCCP, a selective inhibitor of mitochondrial Ca2+ transport in these cells. The nonmitochondrial flux, representing net Ca2+ extrusion across the plasma membrane, has a simple dependence on [Ca2+](i), while the net mitochondrial flux (J(mito)) is biphasic, indicative of Ca+) accumulation during the initial phase of recovery when [Ca2+](i) is high, and net Ca2+ release during later phases of recovery. During each phase, mitochondrial Ca2+ transport has distinct effects on recovery kinetics. J(mito) was separated into components representing mitochondrial Ca2+ uptake and release based on sensitivity to the specific mitochondrial Na(+)/Ca2+ exchange inhibitor, CGP 37157 (CGP). The CGP-resistant (uptake) component of J(mito) increases steeply with [Ca2+](i), as expected for transport by the mitochondrial uniporter. The CGP-sensitive (release) component is inhibited by lowering the intracellular Na(+) concentration and depends on both intra- and extramitochondrial Ca2+ concentration, as expected for the Na(+)/Ca2+ exchanger. Above approximately 400 nM [Ca2+](i), net mitochondrial Ca2+ transport is dominated by uptake and is largely insensitive to CGP. When [Ca2+](i) is approximately 200-300 nM, the net mitochondrial flux is small but represents the sum of much larger uptake and release fluxes that largely cancel. Thus, mitochondrial Ca2+ transport occurs in situ at much lower concentrations than previously thought, and may provide a mechanism for quantitative control of ATP production after brief or low frequency stimuli that raise [Ca(2+)](i) to levels below approximately 500 nM.
Sigma-Aldrich
CGP-37157, ≥98% (HPLC), powder
Pricing and availability is not currently available.