Merck
HPLC Analysis of Sucrose on SeQuant® ZIC-HILIC with Refractive Index Detectio
HPLC Analysis of Sucrose on SeQuant® ZIC-HILIC with Refractive Index Detectio application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate
1.0 mL/min
column temp.
ambient
detector
refractive index
injection
20 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany