Skip to Content
Merck

GF73624013

Tin

foil, 4mm disks, thickness 0.01mm, 97.4%

Synonym(s):

Tin, SN000212

Sign In to View Organizational & Contract Pricing.

Select a Size

10 EA

$1,530.00

$1,530.00


Estimated to ship on20 February 2026Details


Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141745
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Tin, foil, 4mm disks, thickness 0.01mm, 97.4%

InChI

1S/Sn

SMILES string

[Sn]

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

assay

97.4%

form

foil

manufacturer/tradename

Goodfellow 736-240-13

resistivity

11 μΩ-cm, 20°C

diam. × thickness

4 mm × 0.01 mm

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF10510681GF66628996GF92311185
Tin foil, 4mm disks, thickness 0.01mm, 97.4%

GF73624013

Tin

Tin foil, 4mm disks, thickness 0.006mm, 97.4%

GF10510681

Tin

Tin foil, 4mm disks, thickness 0.007mm, 97.4%

GF66628996

Tin

Tin foil, 4mm disks, thickness 0.009mm, 97.4%

GF92311185

Tin

form

foil

form

foil

form

foil

form

foil

assay

97.4%

assay

97.4%

assay

97.4%

assay

≥97.4%

manufacturer/tradename

Goodfellow 736-240-13

manufacturer/tradename

Goodfellow 105-106-81

manufacturer/tradename

Goodfellow 666-289-96

manufacturer/tradename

Goodfellow 923-111-85

density

7.310 g/mL at 25 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

density

7.310 g/mL at 25 °C (lit.)

bp

2270 °C (lit.)

bp

2270 °C (lit.)

bp

2270 °C (lit.)

bp

2270 °C (lit.)

mp

231.9 °C (lit.)

mp

231.9 °C (lit.)

mp

231.9 °C (lit.)

mp

231.9 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianan Liu et al.
Animal reproduction science, 134(3-4), 197-202 (2012-08-28)
Cryopreservation of ovarian tissue has been the only effective way of ex situ conservation of female germplasm in avian species. A novel needle-in-straw (NIS) vitrification method was developed to store tissue in straws instead of cryovials. Fragments of ovarian tissue
J Strang et al.
Addiction (Abingdon, England), 92(6), 673-683 (1997-06-01)
The history of heroin smoking and the subsequent development and spread of 'chasing the dragon' are examined. The first heroin smoking originated in Shanghai in the 1920s and involved use of porcelain bowls and bamboo tubes, thereafter spreading across much
Andrey A Gurtovenko et al.
The Journal of chemical physics, 130(21), 215107-215107 (2009-06-11)
The electrostatic properties of lipid membranes are of profound importance as they are directly associated with membrane potential and, consequently, with numerous membrane-mediated biological phenomena. Here we address a number of methodological issues related to the computation of the electrostatic
K A Winship
Adverse drug reactions and acute poisoning reviews, 7(1), 19-38 (1988-01-01)
Inorganic tin salts are poorly absorbed and rapidly excreted in the faeces; as a result they have a low toxicity. Only about 5 per cent is absorbed from the gastrointestinal tract, widely distributed in the body, then excreted by the
Britta A Jung et al.
Clinical oral implants research, 22(6), 664-668 (2010-11-04)
To evaluate the necessity of three-dimensional imaging (computed tomography [CT]/cone-beam computed tomography [CBCT]) for paramedian insertion of palatal implants. Lateral radiographs and CBCT scans were performed from 18 human skulls. For lateral cephalometry, the nasal floor (right/left) and the oral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service