Skip to Content
Merck

PHL23755

Lepidiline A

new

phyproof® Reference Substance

Synonym(s):

4,5-Dimethyl-1,3-bis(phenylmethyl)-1H-imidazolium chloride, Macaline A

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C19H21ClN2
CAS Number:
Molecular Weight:
312.84
UNSPSC Code:
41116107

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

−20°C

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service