Skip to Content
Merck

SY3428347161

Synthonix Corporation

trans-3-Amino-3-methylcyclobutanol hydrochloride

Synonym(s):

(1r,3r)-3-amino-3-methylcyclobutan-1-ol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product SY3428347161 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
137.61
MDL number:
Product SY3428347161 is not currently sold in your country.

Inchi Code

1S/C5H11NO.ClH/c1-5(6)2-4(7)3-5;/h4,7H,2-3,6H2,1H3;1H/t4-,5-;

Inchi Key

RTAWFZMTTJCICN-BPNVWSNHSA-N

Storage temp.

Refrigerator

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

MSDS

https://synthonix.com/11705.html

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library