Merck
UFC8030
Millipore
Amicon® Ultra Centrifugal Filter, 30 kDa MWCO
greener alternative
sample volume 4 mL, regenerated cellulose membrane, MWCO 30 kDa
Synonym(s):
Centrifugal concentrator,30 kDa MWCO, 4 mL sample volume
Slide 1 of 2
Sign Into View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polyethylene cap
polyethylene liner
polypropylene tube
regenerated cellulose membrane
regenerated cellulose support
styrene-butadiene housing
Quality Level
sterility
non-sterile
product line
Amicon®
packaging
pkg of 24 pkg
pkg of 8 pkg
pkg of 96 pkg
manufacturer/tradename
Amicon® Ultra
sustainability
Greener Alternative Product
parameter
4 mL sample volume
technique(s)
protein extraction: suitable
protein purification: suitable
L
124 mm (4.88 in.)
diam.
17.3 mm
filtration area
3 cm2
volume
4 mL
working volume
≤4 mL
4 mL
pore size
30 kDa MWCO
greener alternative category
shipped in
ambient
General description
Amicon® Ultra 4 mL centrifugal devices with Ultracel® 30,000 Dalton molecular weight cutoff filters provide fast ultrafiltration, and reliable retention of macromolecules. For example, Ovalbumin (45 kDa) at 1 mg/ml initial concentration is retained at >90% .
The vertical design and available membrane surface area provide fast sample processing, high sample recovery (typically greater than 90% of dilute starting solution), and the capability for 80-fold concentration. Solute polarization and subsequent fouling of the membrane are minimized by the vertical design, and a physical deadstop in the filter device prevents spinning to dryness and potential sample loss.
The concentrate is collected from the filter device sample reservoir using a pipetter, while the ultrafiltrate is collected in the provided centrifuge tube. The device can be spun in a swinging bucket or fixed angle rotor. With their vertical membrane design and deadstop, Amicon® Ultra 15 mL filters deliver the highest recovery with short spin times.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner filtration device is made from 60% bio-circular material allocated using the ISCC mass balance approach. Click here for more information.
Application
  • Ultrafiltration with 30,000 Dalton molecular weight cutoff Ultracel® membrane.
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms.
  • Purification of macromolecular components found in tissue culture extracts or cell lysates and protein removal prior to HPLC.
  • Concentration of dilute or pre-purified proteins from column eluents.
  • Desalting, buffer exchange and protein dialysis.
  • Size exclusion and protein fractionation.
Features and Benefits
  • Ultracel® regenerated cellulose membrane with 30 kDa NMWCO delivers retentate recovery of greater than 90%.
  • Vertical membrane reduces concentration polarization for ultra-fast spin times (as fast as 5 minutes).
  • Heat-sealed membrane minimizes downstream extractables.
  • 100% integrity tested for reliable performance.
  • Convenient sample monitoring with translucent housing and volume gradations.
  • Direct pipette sample access eliminates processing step to recover concentrate.
  • High concentration factors of 80–100X.
Physical form
Color Code: Clear with pink cap
Legal Information
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Articles
Filtration methods and membrane filter selection in nanoparticle purification and production.
Explore ultrafiltration membrane principles and benefits for protein sample preparation, including solvent exchange and sample concentration.
Protocols
Biomarkers play an essential role in the drug discovery and development process.
The human plasma proteome is a vast resource for biomarker discovery and investigation.
Questions
1–2 of 2 Questions  
  1. 동일한 단백질에 대해서 정제 후 필터 재사용 가능한가요?
    1 answer
    1. This product is intended for single use only. Repeated use may impact filter integrity resulting poor filtration.
      Helpful?
  2. Do you have any sterile spin filter options which have the same specification as the one selected?
    1 answer
    1. Unfortunately, sterile versions of these concentrator devices are not available. The Amicon filters cannot be sterilized, but can be "sanitized" with a wash in 70 % ethanol followed by 2-3 flushes with sterile water.
      It is recommended to filter sterilize solutions after processing with the Amicon.
      Helpful?
Reviews
No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service