Inchi Code
1S/C5H6ClNS/c6-2-1-5-3-8-4-7-5/h3-4H,1-2H2
Inchi Key
SJKXFYXCQCXHIH-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 4-(2-Chloroethyl)thiazole
hstatements
Precautionary Statements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

