Product BDPH9BCD047E is not currently sold in your country.
Inchi Code
1S/C8H8Cl2N2/c9-7-5-3-1-2-4-6(5)8(10)12-11-7/h1-4H2
Inchi Key
KLFNXIXNWZZRLE-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Country of Origin
CN
Physical Form
Solid
Storage temp.
Inert atmosphere, 2-8°C
Still not finding the right product?
Explore all of our products under 1,4-Dichloro-5,6,7,8-tetrahydrophthalazine
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
