Skip to Content
Merck

BDPH9BCD1FCA

Bide Pharmatech

8-Methyldibenzo[b,f][1,4]thiazepin-11(10H)-one

Synonym(s):

8-methyldibenzo[b,f][1,4]thiazepin-11(10H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H11O1N1S1
CAS Number:
Molecular Weight:
241.31
MDL number:
Product BDPH9BCD1FCA is not currently sold in your country.


Inchi Code

1S/C14H11NOS/c1-9-6-7-13-11(8-9)15-14(16)10-4-2-3-5-12(10)17-13/h2-8H,1H3,(H,15,16)

Inchi Key

JNUXHEWGPQCJRD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library