Skip to Content
Merck

BDPH9C021C9B

Bide Pharmatech

(4R,5R)-4,5-Diphenyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole

Synonym(s):

(4R,5R)-4,5-diphenyl-2-(6-phenylpyridin-2-yl)-4,5-dihydrooxazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C26H20O1N2
CAS Number:
Molecular Weight:
376.46
MDL number:
Product BDPH9C021C9B is not currently sold in your country.


Inchi Code

1S/C26H20N2O/c1-4-11-19(12-5-1)22-17-10-18-23(27-22)26-28-24(20-13-6-2-7-14-20)25(29-26)21-15-8-3-9-16-21/h1-18,24-25H/t24-,25-/m1/s1

Inchi Key

GEGIOXAHQGYSFZ-JWQCQUIFSA-N

Shipping Temperature

Normal

Purity

98%99%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library