Skip to Content
Merck

BDPH9C021ED8

Bide Pharmatech

2-(1-Methyl-1H-pyrazol-3-yl)phenol

Synonym(s):

2-(1-methyl-1H-pyrazol-3-yl)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O1N2
CAS Number:
Molecular Weight:
174.2
MDL number:
Product BDPH9C021ED8 is not currently sold in your country.


Inchi Code

1S/C10H10N2O/c1-12-7-6-9(11-12)8-4-2-3-5-10(8)13/h2-7,13H,1H3

Inchi Key

NYHQEXCUPFTZHJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?

Explore all of our products under 2-(1-Methyl-1H-pyrazol-3-yl)phenol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library