Skip to Content
Merck

PG9H9BB386C0

PrecisePEG

Fmoc-pSar10-OH

Synonym(s):

2-(2-{2-[2-(2-{2-[2-(2-{2-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-N-methylacetamido]-N-methylacetamido}-N-methylacetamido)-N-methylacetamido]-N-methylacetamido}-N-methylacetamido)-N-methylacetamido]-N-methylacetamido}-N-methylacetamido)acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C45H62O13N10
CAS Number:
Molecular Weight:
951.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C45H62N10O13/c1-46(20-36(57)48(3)22-38(59)50(5)24-40(61)52(7)26-42(63)54(9)28-44(65)66)35(56)19-47(2)37(58)21-49(4)39(60)23-51(6)41(62)25-53(8)43(64)27-55(10)45(67)68-29-34-32-17-13-11-15-30(32)31-16-12-14-18-33(31)34/h11-18,34H,19-29H2,1-10H3,(H,65,66)

Inchi Key

LWJFVEBBBTXGOQ-UHFFFAOYSA-N

Shipping Temperature

ambient

Purity

96%

Storage temp.

-20

Country of Origin

CN

Physical Form

Solid



hstatements

Precautionary Statements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library