
Dilithium tetrabromonickelate(II) solution

Synonym(s):
Lithium tetrabromonickelate(II) solution, di-Lithium tetrabromonickelate(II)
Linear Formula:
Li2NiBr4
CAS Number:
Molecular Weight:
392.19
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing