Merck
  • Influence of tobacco smoking on transferrin sialylation during pregnancy in smoking and non-smoking women with iron deficiency
Influence of tobacco smoking on transferrin sialylation during pregnancy in smoking and non-smoking women with iron deficiency
Environmental Toxicology and Pharmacology (2016)
Wrzesniak M, et al.