Merck
  • Production of neutralizing antibodies against the secreted Clostridium chauvoei toxin A (CctA) upon blackleg vaccination.
Production of neutralizing antibodies against the secreted Clostridium chauvoei toxin A (CctA) upon blackleg vaccination.
Anaerobe (2019-02-17)
Pamela Nicholson, Julia Furrer, Michael Hässig, Christian Strauss, Manfred Heller, Sophie Braga-Lagache, Joachim Frey
ABSTRACT
Clostridium chauvoei is the etiologic agent of blackleg in cattle, inducing fever, severe myonecrosis, oedemic lesions and ultimately death of infected animals. The pathogen often results in such rapid death that antibiotic therapy is futile and thus vaccination is the only efficient strategy in order to control the disease. The β-barrel pore forming leucocidin Clostridium chauvoei toxin A (CctA) is one of the best characterised toxins of C. chauvoei and has been shown to be an important virulence factor. It has been reported to induce protective immunity and is conserved across C. chauvoei strains collected from diverse geographical locations for more than 50 years. The aim of this study was to identify the location of the CctA toxin during liquid culture fermentation and to use CctA to develop an in vitro assay to replace the current guinea pig challenge assay for vaccine potency in standard batch release procedures. We report that CctA is fully secreted in C. chauvoei culture and show that it is found abundantly in the supernatant of liquid cultures. Sera from cattle vaccinated with a commercial blackleg vaccine revealed strong haemolysin-neutralizing activity against recombinant CctA which reached titres of 1000 times 28 days post-vaccination. Similarly, guinea pig sera from an official potency control test reached titres of 600 times 14 days post-vaccination. In contrast, ELISA was not able to specifically measure anti-CctA antibodies in cattle serum due to strong cross-reactions with antibodies against other proteins present pre-vaccination. We conclude that haemolysin-neutralizing antibodies are a valuable measurement for protective immunity against blackleg and have the potential to be a suitable replacement of the guinea pig challenge potency test, which would forego the unnecessary challenge of laboratory animals.
Sigma-Aldrich
Monoclonal Anti-Bovine IgG−Alkaline Phosphatase antibody produced in mouse, clone BG-18, purified immunoglobulin, buffered aqueous glycerol solution
Pricing and availability is not currently available.