Merck
  • Combinatorial Sensor Design in Caulobacter crescentus for Selective Environmental Uranium Detection.
Combinatorial Sensor Design in Caulobacter crescentus for Selective Environmental Uranium Detection.
ACS synthetic biology (2019-03-22)
Dan M Park, Michael J Taffet
ABSTRACT
The ability to detect uranium (U) through environmental monitoring is of critical importance for informing water resource protection and nonproliferation efforts. While technologies exist for environmental U detection, wide-area environmental monitoring, i.e. sampling coverage over large areas not known to possess U contamination, remains a challenging prospect that necessitates the development of novel detection approaches. Herein, we describe the development of a whole-cell U sensor by integrating two functionally independent, native U-responsive two-component signaling systems (TCS), UzcRS and UrpRS, within an AND gate circuit in the bacterium Caulobacter crescentus. Through leverage of the distinct but imperfect selectivity profiles of both TCS, this combinatorial approach enabled greater selectivity relative to a prior biosensor developed with UzcRS alone; no cross-reactivity was observed with most common environmental metals (e.g, Fe, As, Cu, Ca, Mg, Cd, Cr, Al) or the U decay-chain product Th, and the selectivity against Zn and Pb was significantly improved. In addition, integration of the UzcRS signal amplifier protein UzcY within the AND gate circuit further enhanced overall sensitivity and selectivity for U. The functionality of the sensor in an environmental context was confirmed by detection of U concentrations as low as 1 μM in groundwater samples. The results highlight the value of a combinatorial approach for constructing whole-cell sensors for the selective detection of analytes for which there are no known evolved regulators.