Merck
  • Unravelling the phytonutrients and antioxidant properties of European Vicia faba L. seeds.
Unravelling the phytonutrients and antioxidant properties of European Vicia faba L. seeds.
Food research international (Ottawa, Ont.) (2019-02-06)
Inês M Valente, Ana R J Cabrita, Nertila Malushi, Hugo M Oliveira, Lumturi Papa, José A Rodrigues, António J M Fonseca, Margarida R G Maia
ABSTRACT
For the first time, seven European varieties of Vicia faba L. seeds were investigated for (1) their phytonutrients profile by HPLC-DAD-MS/MS, (2) total phenolic content (TPC), and (3) antioxidant capacity (DPPH and FRAP assays). A wide range of compounds were identified, namely alkaloids, organic acids, terpenoids, jasmonates, and phenolics; these two latter being the most abundant. TPC ranged between 2.62 and 4.3 mg (gallic acid equivalent) g-1 dry weight, for V. faba major variety Belshi and V. faba minor variety Bauska, respectively. The DPPH radical scavenging capacity showed poor correlation (r = 0.550, P = .041) with TPC, suggesting the presence of other antioxidant sources than phenolics. Still, FRAP was positively correlated with TPC (r = 0.709, P < .01) and DPPH (r = 0.819, P < .01). These results elucidated the phytonutrients and antioxidant properties of V. faba L. seeds as functional food sources.