Merck
  • EGF-GH Axis in Rat Steatotic and Non-steatotic Liver Transplantation From Brain-dead Donors.
EGF-GH Axis in Rat Steatotic and Non-steatotic Liver Transplantation From Brain-dead Donors.
Transplantation (2019-06-27)
Ana I Álvarez-Mercado, Elsa Negrete-Sánchez, José Gulfo, Cindy G Ávalos de León, Araní Casillas-Ramírez, María Eugenia Cornide-Petronio, Esther Bujaldon, Floriana Rotondo, Jordi Gracia-Sancho, Mónica B Jiménez-Castro, Carmen Peralta
ABSTRACT
We evaluated the potential dysfunction caused by changes in growth hormone (GH) levels after brain death (BD), and the effects of modulating GH through exogenous epidermal growth factor (EGF) in steatotic and nonsteatotic grafts. Steatotic and nonsteatotic grafts from non-BD and BD rat donors were cold stored for 6 hours and transplanted to live rats. Administration of GH and EGF and their underlying mechanisms were characterized in recipients of steatotic and nonsteatotic grafts from BD donors maintained normotensive during the 6 hours before donation. Circulating and hepatic GH and EGF levels, hepatic damage, and regeneration parameters were evaluated. Recipient survival was monitored for 14 days. Somatostatin, ghrelin, and GH-releasing hormones that regulate GH secretion from the anterior pituitary were determined. The survival signaling pathway phosphoinositide-3-kinase/protein kinase B that regulates inflammation (suppressors of cytokine signaling, high-mobility group protein B1, oxidative stress, and neutrophil accumulation) was evaluated. BD reduced circulating GH and increased GH levels only in steatotic livers. GH administration exacerbated adverse BD-associated effects in both types of graft. Exogenous EGF reduced GH in steatotic livers, thus activating cell proliferation and survival signaling pathways, ultimately reducing injury and inflammation. However, EGF increased GH in nonsteatotic grafts, which exacerbated damage. The benefits of EGF for steatotic grafts were associated with increased levels of somatostatin, a GH inhibitor, whereas the deleterious effect on nonsteatotic grafts was exerted through increased amounts of ghrelin, a GH stimulator. GH treatment is not appropriate in rat liver transplant from BD donors, whereas EGF (throughout GH inhibition) protects only in steatotic grafts.