Merck
  • Highly spectral dependent enhancement of upconversion emission with sputtered gold island films.
Highly spectral dependent enhancement of upconversion emission with sputtered gold island films.
Chemical communications (Cambridge, England) (2010-11-17)
Hua Zhang, Di Xu, Yu Huang, Xiangfeng Duan
ABSTRACT
We report a five-fold overall enhancement of upconversion emission in NaYF(4) : Yb/Er nanocrystals when coupled with gold island films. Spectroscopic studies show that the enhancement factors are highly dependent on the exact spectral positions and excitation power density, with a largest enhancement factor of more than 12 observed at selected spectral positions, which may be attributed to different upconversion processes involved.
Sigma-Aldrich
Sodium yttrium fluoride, ytterbium and erbium doped, green up-conversion phosphor, 99% trace metals basis