Merck
  • Evidence for antimanic efficacy of glycogen synthase kinase-3 (GSK3) inhibitors in a strain-specific model of acute mania.
Evidence for antimanic efficacy of glycogen synthase kinase-3 (GSK3) inhibitors in a strain-specific model of acute mania.
The international journal of neuropsychopharmacology (2011-01-07)
Mikhail Kalinichev, Lee A Dawson
ABSTRACT
There is a growing body of evidence suggesting that animal models can be developed to probe the specific domains of bipolar disorder (BD) using the endophenotype approach. Here we tested clinically active antimanic drugs to validate amphetamine-induced hyperactivity in Black Swiss mice as a putative model of the manic phase of BD. We also co-administered a mood stabilizer and an atypical antipsychotic drug in a manner akin to the clinical treatment regimens. Since lithium has been shown to potentially act through glycogen synthase kinase-3 (GSK3) inhibition, we evaluated the efficacy of selective GSK3 inhibitors in this model. Habituated animals were pretreated with a compound of interest before being challenged with amphetamine (2.0 mg/kg) and returned to activity cages for an additional 1.5 h. We tested lithium, sodium valproate, carbamazepine, olanzapine, ziprasidone as well as co-administered lithium and olanzapine at sub-efficacious doses. The GSK3 inhibitors tested included indirubin, alsterpaullone, TDZD-8, AR-A014418, SB-216763, and SB-627772. All mood stabilizers and antipsychotic drugs reduced hyperactivity without affecting spontaneous locomotion. While subactive doses of lithium and olanzapine were without effect, their co-administration produced robust reductions in hyperactivity. All GSK3 inhibitors were active in the model, producing selective inhibition of rearing hyperactivity. These data support the predictive validity of the model for the acute manic phase of BD and may have utility as an in-vivo model for identifying novel antimanic therapeutics.
Sigma-Aldrich
Alsterpaullone, ≥98% (HPLC), powder
Pricing and availability is not currently available.