Merck
  • Application of solvent microextraction to the analysis of nitroaromatic explosives in water samples.
Application of solvent microextraction to the analysis of nitroaromatic explosives in water samples.
Journal of chromatography. A (2001-02-24)
E Psillakis, N Kalogerakis
ABSTRACT
The application of solvent microextraction to the analysis of nitroaromatic explosives is presented. Extraction of 11 nitroaromatics was achieved by suspending 1 microl of organic solvent to the tip of a microsyringe in a stirred aqueous solution. Parameters such as extraction solvent, stirring rate, salt concentration and sampling time were studied and optimized. The limits of detection using bench-top quadrupole mass spectrometry and short extraction times (15 min) were found to be between 0.08 and 1.3 microg/l and the relative standard deviations ranged between 4.3 and 9.8%. Although precision and accuracy of quantification of the method are still needed, solvent microextraction proved to be a fast, simple and inexpensive tool for preconcentration and matrix isolation of nitroaromatics on a microscale.
Product Description
Supelco
2,3-Dinitrotoluene, analytical standard