Merck
  • Activity of the Diaminopyrimidine AR-709 against recently collected multidrug-resistant isolates of invasive Streptococcus pneumoniae from North America.
Activity of the Diaminopyrimidine AR-709 against recently collected multidrug-resistant isolates of invasive Streptococcus pneumoniae from North America.
Antimicrobial agents and chemotherapy (2008-01-09)
Roseanne A Ressner, Matthew R Moore, James H Jorgensen
ABSTRACT
Broth microdilution was used to determine the MICs of AR-709 and comparator antimicrobial agents for 224 invasive multidrug-resistant isolates of Streptococcus pneumoniae. AR-709 was highly active, with a MIC 50 of 0.25 microg/ml, a MIC 90 of 0.5 microg/ml, and a range of <or=0.008 microg/ml to 1 microg/ml.
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.