Merck
  • Insulin Receptor Substrate p53 Ameliorates High-Glucose-Induced Activation of NF-κB and Impaired Mobility of HUVECs.
Insulin Receptor Substrate p53 Ameliorates High-Glucose-Induced Activation of NF-κB and Impaired Mobility of HUVECs.
BioMed research international (2021-01-29)
Fen Liu, Yubin Chen, Shi Zhao, Mei Li, Fanyan Luo, Can-E Tang
ABSTRACT
Diabetes-related macrovascular and microvascular complications lead to poor prognosis. Insulin receptor substrate p53 (IRSp53) is known to act as a substrate for the insulin receptor tyrosine kinase, but its role in endothelial dysfunction remains unclear. Human umbilical vein endothelial cells (HUVECs) treated with D-glucose at different concentrations and a streptozocin-induced rat diabetes mellitus (DM) model were used to investigate the effects of hyperglycemia on the expression levels of IRSp53 and galectin-3 (gal-3) and the inflammatory state and mobility of HUVECs. Thereafter, IRSp53-overexpressing HUVECs and IRSp53-knockdown HUVECs were established using IRSp53-overexpressing lentivirus or IRSp53-siRNA to explore the role of IRSp53 in the HUVEC inflammatory state and HUVEC mobility. D-glucose at high concentration (HG) and hyperglycemia were found to induce downregulation of IRSp53 and upregulation of gal-3 in vitro and in vivo. Treatment with HG resulted in activation of NF-κB in HUVECs and impaired HUVEC mobility. Insulin restored HG-induced changes in the expression levels of IRSp53 and gal-3 in HUVECs and protected the cells from NF-κB activation and impaired mobility. Overexpression of IRSp53 inhibited the activation of NF-κB in HUVECs and strengthened HUVEC migration. Knockdown of IRSp53 facilitated the activation of NF-κB in HUVECs and decreased HUVEC migration. However, neither overexpression nor knockdown of IRSp53 altered the effects of insulin on HG-induced detrimental changes in HUVECs. HG and hyperglycemia resulted in downregulation of IRSp53 in vitro and in vivo. IRSp53 is concluded to inhibit the activation of NF-κB in HUVECs and to strengthen HUVEC migration.
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Insulin human, meets USP testing specifications
Sigma-Aldrich
2-Phenylindole, technical grade, 95%