Merck
  • 4,5,6,7-Tetrahydroisothiazolo[5,4-c]pyridin-3-ol and related analogues of THIP. Synthesis and biological activity.
4,5,6,7-Tetrahydroisothiazolo[5,4-c]pyridin-3-ol and related analogues of THIP. Synthesis and biological activity.
Journal of medicinal chemistry (1983-06-01)
P Krogsgaard-Larsen, H Mikkelsen, P Jacobsen, E Falch, D R Curtis, M J Peet, J D Leah
ABSTRACT
The thio analogues of the GABA (gamma-aminobutyric acid) agonist THIP (4,5,6,7-tetrahydroisoxazolo[5,4-c]pyridin-3-ol), the GABA uptake inhibitor THPO (4,5,6,7-tetrahydroisoxazolo[4,5-c]pyridin-3-ol), and the glycine antagonist THAZ (5,6,7,8-tetrahydro-4H-isoxazolo[4,5-d]azepin-3-ol) have been synthesized and tested biologically on single neurons in the cat spinal cord and in vitro by using synaptic membrane preparations obtained from rat brains. In contrast to THIP, thio-THIP (4,5,6,7-tetrahydroisothiazolo[5,4-c]pyridin-3-ol, 5) was only a weak GABA agonist. Thio-THPO (4,5,6,7-tetrahydroisothiazolo[4,5-c]pyridin-3-ol, 10) was slightly weaker than THPO as an inhibitor of GABA uptake in vitro, and these two compounds were approximately equipotent in enhancing the inhibition of the firing of cat spinal neurons by GABA. Like THAZ and structurally related bicyclic isoxazole zwitterions, thio-THAZ (5,6,7,8-tetrahydro-4H-isothiazolo[4,5-d]azepin-3-ol, 15) was an antagonist at glycine receptors on cat spinal neurons. The I/U ratios, which reflect the ability of neutral amino acids to penetrate the blood-brain barrier (BBB), were calculated for 5 (I/U = 16), 10 (63), and 15 (200). These low I/U ratios, compared with the findings that THIP (I/U = 500 or 1500) and THPO (I/U = 2500) enter the brain after systemic administration, suggest that the thio analogues may penetrate the BBB very easily.
Sigma-Aldrich
Gaboxadol hydrochloride, solid, ≥98% (HPLC)
Pricing and availability is not currently available.