Merck
  • Prolyl endopeptidase disruption reduces hepatic inflammation and oxidative stress in methionine-choline-deficient diet-induced steatohepatitis.
Prolyl endopeptidase disruption reduces hepatic inflammation and oxidative stress in methionine-choline-deficient diet-induced steatohepatitis.
Life sciences (2021-02-01)
Jianbin Zhang, Daixi Jiang, Shuangzhe Lin, Yuqing Cheng, Jiaxing Pan, Wenjin Ding, Yuanwen Chen, Jiangao Fan
ABSTRACT
Prolyl endopeptidase (PREP) is a serine endopeptidase widely distributed in the body, and accumulated evidence suggests that PREP participates in inflammation and oxidative stress. Here, we explored the effect of PREP gene disruption on hepatic inflammation and oxidative stress status in a methionine-choline-deficient (MCD)-induced nonalcoholic steatohepatitis (NASH) model. PREP gene disruption (PREPgt) mice and wild-type (WT) littermates were placed on a control or an MCD diet for 4 weeks, respectively. The liver histopathological analysis and the number of inflammatory cells were determined by hematoxylin-eosin (HE) and immunohistochemical staining. Inflammation-associated genes and cytokine levels in liver tissue were evaluated by quantitative PCR and ELISA. The levels of P53, Sesn2, Nrf2, HO-1, and oxidative stress indicators in mice and the palmitic acid (PA)-treated human hepatocellular carcinoma cells (HepG2) were examined by immunoblotting and commercially available kits, respectively. We found that PREP expression was upregulated in the MCD-induced NASH model. In addition, PREP disruption alleviated MCD-induced hepatic inflammation accompanied by diminished infiltration of inflammatory cells and secretion of inflammatory mediators. More importantly, the results of this study indicate that targeting PREP can improve oxidative stress status in the liver of MCD-diet mice and PA-exposed HepG2 cells. The effect is most likely mediated by the activation of P53 and its downstream signaling pathways (Sesn2/Nrf2/HO-1). Our results showed that PREP disruption (or inhibition) could decrease oxidative stress and inflammation and improve liver function, indicating that targeting PREP might be a new potential therapeutic option for NAFLD/NASH.
Sigma-Aldrich
KYP-2047, ≥95% (HPLC)
Pricing and availability is not currently available.