Merck
  • [Effect of long chain non-coding RNA H19 on the migration and invasion of oral cancer cells and its molecular mechanism].
[Effect of long chain non-coding RNA H19 on the migration and invasion of oral cancer cells and its molecular mechanism].
Hua xi kou qiang yi xue za zhi = Huaxi kouqiang yixue zazhi = West China journal of stomatology (2019-09-13)
Jun-Fang Zhao, Zhi-An Zha, Wei-Hong Xie, Hai-Bin Wang, Xin-Ming Li, Qiang Sun, Ming-Lei Sun
ABSTRACT
To investigate the effect of the long chain non-coding RNA H19 (lncRNA H19) on the invasion and migration of oral cancer cells and its related molecular mechanism. The expression levels of lncRNA H19, miR-107, and cyclin-dependent kinase 6 (CDK6) in the immortalized oral epithelial cell line HIOEC and the oral cancer cell line CAL27 were detected by real-time quantitative polymerase chain reaction. CAL27 cells were transfected with siRNA H19, miR-107 mimics, pcDNA H19, or anti-miR-107, and the effects of H19 and miR-107 on the invasion and migration of cells were examined via Transwell assay. The TargetScan database predicted the targeting of H19, miR-107, and CDK6. Double luciferase reporter gene assay was performed to detect interactions among H19, miR-107, and CDK6. Western blot analysis was conducted to examine the effects of H19 and miR-107 on the protein level of the target gene CDK6. Compared with that in HIOEC cells, the expression of H19 was significantly increased in CAL27 cells (P<0.05). After transfection with siRNA H19, the expression of H19 decreased, and the invasion and migration ability of CAL27 cells were inhibited (P<0.05). H19 could bind specifically to the 3'-UTR of miR-107 to modulate the expression of miR-107. Compared with that in HIOEC cells, the expression of miR-107 significantly decreased in CAL27 cells (P<0.05). The expression of miR-107 increased after transfection with siRNA H19, and anti-mir-107 co-transfection could promote the invasion and migration ability of siRNA H19 in CAL27 cells (P<0.05). Compared with that in HIOEC cells, CDK6 expression significantly increased in CAL27 cells (P<0.05), and the expression level of the gene was coregulated by H19 and miR-107 (P<0.05). lncRNA H19 plays an important role in the development of oral cancer. It can regulate the invasion and migration of oral cancer cells by targeting the miR-107/CDK6 signaling axis.
Sigma-Aldrich
Imprint® RNA Immunoprecipitation Kit, High-capacity Protein A magnetic beads for successful RNA Immunoprecipitation,suitable for use with mRNA and microRNA
Pricing and availability is not currently available.