Merck
  • Self-selection in the self-assembly of isomeric supramolecular squares from unsymmetrical bis(4-pyridyl)acetylene ligands.
Self-selection in the self-assembly of isomeric supramolecular squares from unsymmetrical bis(4-pyridyl)acetylene ligands.
The Journal of organic chemistry (2008-08-08)
Liang Zhao, Brian H Northrop, Yao-Rong Zheng, Hai-Bo Yang, Hyo Jin Lee, Young Min Lee, Joo Yeon Park, Ki-Whan Chi, Peter J Stang
ABSTRACT
A new approach wherein steric interactions between substituents of unsymmetrical bis(4-pyridyl)acetylene ligands dictate the self-selection of single isomers of [4 + 4] self-assembled squares is presented. Each [4 + 4] self-assembly is characterized by multinuclear (31)P and (1)H NMR spectroscopies and electrospray ionization mass spectrometry. NMR spectroscopic studies are used to provide a means of evaluating the efficiency of bulky substituents at proximal or remote positions relative to the Pt-N bonding motif to direct self-selection. Molecular modeling using the MMFF force field is utilized to determine the relative energy of different isomers of each assembly, and modeling results reasonably explain the trend in self-selectivity with varying pyridyl substitution.
Sigma-Aldrich
2,4-Dichloro-3-iodopyridine, 97%
Sigma-Aldrich
2,6-Dichloro-3-iodopyridine, 97%
Sigma-Aldrich
2,3-Dichloro-4-iodopyridine, 97%
Sigma-Aldrich
2,6-Dichloro-4-iodopyridine, 97%
Sigma-Aldrich
3,5-Dichloro-4-iodopyridine, 97%