Merck
  • EBP1 promotes the malignant biological behaviors of kidney renal clear cell carcinoma through activation of p38/HIF-1α signaling pathway.
EBP1 promotes the malignant biological behaviors of kidney renal clear cell carcinoma through activation of p38/HIF-1α signaling pathway.
Cancer cell international (2024-07-26)
Huan Meng, Shuxia Cao, Shengri Tian, Jiaqi Huo, Xiangdan Li, Dongyuan Xu, Lan Liu
ABSTRACT
Kidney Renal Clear Cell Carcinoma (KIRC) is a common malignant tumor of the urinary system, and its incidence is increasing. ERBB3 binding protein (EBP1) is upregulated in various cancers. However, the connection between EBP1 and KIRC has not been reported. The expression of EBP1 in normal kidney tissue and KIRC tissue was analyzed through database and tissue microarray. EBP1 was knocked down in KIRC cell lines, and its impact on KIRC proliferation was assessed through CCK-8, soft agar assay, and flow cytometry. Scratch and transwell assays were used to evaluate the influence of EBP1 on KIRC invasion and migration. Nude mice tumor experiment were conducted to examine the effect of EBP1 on tumor tissue. Database analysis explored potential pathways involving EBP1, and validation was performed through Western blot experiments and p38 inhibitor. EBP1 is upregulated in KIRC and significantly correlates with clinical staging, pathological grading, and lymph node metastasis in patients. The mechanism research showed that knocking down EBP1 inhibited KIRC proliferation, invasion, and migration and inhibited p38 phosphorylation and the expression of hypoxia-inducible factor-1α (HIF-1α) in KIRC. p-38 inhibitor (SB203580) inhibits p38 phosphorylation and HIF-1α expression and suppresses cell viability in a concentration-dependent manner, but has no effect on EBP1 expression. HEK 293T cells overexpressing EBP1 showed increased expression of phosphorylated p38 and HIF-1α and enhanced cell viability, however, SB203580 inhibited this effect of EBP1. EBP1 may promote the occurrence and development of KIRC by regulating the expression of p38/HIF-1α signaling pathway.