Merck
  • Studies on the nonmevalonate terpene biosynthetic pathway: metabolic role of IspH (LytB) protein.
Studies on the nonmevalonate terpene biosynthetic pathway: metabolic role of IspH (LytB) protein.
Proceedings of the National Academy of Sciences of the United States of America (2002-01-31)
Felix Rohdich, Stefan Hecht, Katrin Gärtner, Petra Adam, Cornelia Krieger, Sabine Amslinger, Duilio Arigoni, Adelbert Bacher, Wolfgang Eisenreich
ABSTRACT
Isopentenyl diphosphate and dimethylallyl diphosphate serve as the universal precursors for the biosynthesis of terpenes. Although their biosynthesis by means of mevalonate has been studied in detail, a second biosynthetic pathway for their formation by means of 1-deoxy-D-xylulose 5-phosphate has been discovered only recently in plants and certain eubacteria. Earlier in vivo experiments with recombinant Escherichia coli strains showed that exogenous 1-deoxy-D-xylulose can be converted into 1-hydroxy-2-methyl-2-(E)-butenyl 4-diphosphate by the consecutive action of enzymes specified by the xylB and ispCDEFG genes. This article describes the transformation of exogenous [U-(13)C(5)]1-deoxy-D-xylulose into a 5:1 mixture of [U-(13)C(5)]isopentenyl diphosphate and [U-(13)C(5)]dimethylallyl diphosphate by an E. coli strain engineered for the expression of the ispH (lytB) gene in addition to recombinant xylB and ispCDEFG genes.
Sigma-Aldrich
(E)-1-Hydroxy-2-methyl-2-butenyl 4-pyrophosphate lithium salt, ≥95.0% (TLC)
Pricing and availability is not currently available.