Merck
  • Role for lysosomal phospholipase A2 in iNKT cell-mediated CD1d recognition.
Role for lysosomal phospholipase A2 in iNKT cell-mediated CD1d recognition.
Proceedings of the National Academy of Sciences of the United States of America (2013-03-16)
Crina Paduraru, Jelena S Bezbradica, Amit Kunte, Robert Kelly, James A Shayman, Natacha Veerapen, Liam R Cox, Gurdyal S Besra, Peter Cresswell
ABSTRACT
Invariant natural killer T (iNKT) cells recognize self lipid antigens presented by CD1d molecules. The nature of the self-antigens involved in the development and maturation of iNKT cells is poorly defined. Lysophospholipids are self-antigens presented by CD1d that are generated through the action of phospholipases A1 and A2. Lysosomal phospholipase A2 (LPLA2, group XV phospholipase A2) resides in the endocytic system, the main site where CD1d antigen acquisition occurs, suggesting that it could be particularly important in CD1d function. We find that Lpla2(-/-) mice show a decrease in iNKT cell numbers that is neither the result of a general effect on the development of lymphocyte populations nor of effects on CD1d expression. However, endogenous lipid antigen presentation by CD1d is reduced in the absence of LPLA2. Our data suggest that LPLA2 plays a role in the generation of CD1d complexes with thymic lipids required for the normal selection and maturation of iNKT cells.
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein