Merck
  • [Isolation and purification of Mn-peroxidase from Azospirillum brasilense Sp245].
[Isolation and purification of Mn-peroxidase from Azospirillum brasilense Sp245].
Prikladnaia biokhimiia i mikrobiologiia (2012-05-10)
M A Kupriashina, N Iu Selivanov, V E Nikitina
ABSTRACT
Homogenous Mn-peroxidase of a 26-fold purity grade was isolated from a culture of Azospirillum brasilense Sp245 cultivated on a medium containing 0.1 mM pyrocatechol. The molecular weight of the enzyme is 43 kD as revealed by electrophoresis in SDS-PAAG. It was shown that the use of pyrocatechol and 2,2'-azino-bis(3-ethylbenzotiazoline-6-sulfonate) at concentrations of 0.1 and I mM as inductors increased the Mn-peroxidase activity by a factor of 3.
Sigma-Aldrich
Manganese peroxidase from white-rot fungus (Phanerochaete chrysosporium), powder, light brown, ≥10 U/g
Sigma-Aldrich
Manganese Peroxidase from Nematoloma frowardii, ≥4.2 U/mg